4-((4-Chlorophenoxy)methyl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

67886-500MG 67886-1G 67886-5G
77336-270EA 384.19 USD
77336-270 77357-254 77381-172
4-((4-Chlorophenoxy)methyl)benzaldehyde ≥97%
4-((4-Chlorophenoxy)methyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₄H₁₁ClO₂
Storage Temperature: Ambient
MDL Number: MFCD34763052
CAS Number: 1442085-13-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR