(4-Chloro-2-fluoro-5-methoxyphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

67876-500MG 67876-1G 67876-5G 67876-25G
77333-040EA 108.15 USD
77333-040 77343-198 77369-070 77381-168
(4-Chloro-2-fluoro-5-methoxyphenyl)(methyl)sulfane ≥97%
(4-Chloro-2-fluoro-5-methoxyphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₈H₈ClFOS
MW: 206.665 g/mol
Storage Temperature: Ambient
CAS Number: 2705796-07-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR