(2-Bromo-6-fluoro-3-methylphenyl)trimethylsilane ≥97%

Supplier: AOBChem USA

Warning

67873-500MG 67873-1G 67873-5G 67873-25G
77333-036EA 95.79 USD
77333-036 77342-624 77369-066 77381-164
(2-Bromo-6-fluoro-3-methylphenyl)trimethylsilane ≥97%
(2-Bromo-6-fluoro-3-methylphenyl)trimethylsilane

Precaución: for Research Use Only

Formula: C₁₀H₁₄BrFSi
MW: 261.206 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763050
CAS Number: 2807469-91-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR