(6-Bromo-2-fluoro-3-methylphenyl)trimethylsilane ≥97%

Supplier: AOBChem USA

Warning

67872-1G 67872-5G 67872-25G
77341-436EA 85.49 USD
77341-436 77369-064 77381-162
(6-Bromo-2-fluoro-3-methylphenyl)trimethylsilane ≥97%
(6-Bromo-2-fluoro-3-methylphenyl)trimethylsilane

Precaución: for Research Use Only

Formula: C₁₀H₁₄BrFSi
MW: 261.206 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763049
CAS Number: 2807463-82-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR