(4-Bromo-2,3-difluorophenyl)trimethylsilane ≥97%

Supplier: AOBChem USA

Warning

67871-500MG 67871-1G 67871-5G 67871-25G
77333-034EA 139.05 USD
77333-034 77344-680 77369-062 77390-182
(4-Bromo-2,3-difluorophenyl)trimethylsilane ≥97%
(4-Bromo-2,3-difluorophenyl)trimethylsilane

Precaución: for Research Use Only

Formula: C₉H₁₁BrF₂Si
Storage Temperature: Ambient
MDL Number: MFCD30178762
CAS Number: 1007571-40-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR