2-Bromo-4-ethyl-4'-fluoro-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

67866-500MG 67866-1G 67866-5G 67866-25G
77333-032EA 130.81 USD
77333-032 77344-354 77369-060 77381-160
2-Bromo-4-ethyl-4'-fluoro-1,1'-biphenyl ≥97%
2-Bromo-4-ethyl-4'-fluoro-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₂BrF
MW: 279.147 g/mol
Storage Temperature: Ambient
CAS Number: 2918958-06-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR