2-Chloro-4-fluoro-3-isopropoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

67857-500MG 67857-1G 67857-5G
77337-212EA 408.91 USD
77337-212 77357-250 77381-156
2-Chloro-4-fluoro-3-isopropoxybenzoic acid ≥97%
2-Chloro-4-fluoro-3-isopropoxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₀ClFO₃
Storage Temperature: Ambient
MDL Number: MFCD34763046
CAS Number: 2807441-56-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR