5-Bromo-2-fluoro-3-methoxy-4'-(trifluoromethyl)-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

67847-500MG 67847-1G 67847-5G 67847-25G
77333-022EA 281.19 USD
77333-022 77357-242 77369-052 77390-174
5-Bromo-2-fluoro-3-methoxy-4'-(trifluoromethyl)-1,1'-biphenyl ≥97%
5-Bromo-2-fluoro-3-methoxy-4'-(trifluoromethyl)-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₉BrF₄O
MW: 349.118 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763042
CAS Number: 2901106-74-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR