5-Bromo-2-fluoro-3,4'-dimethoxy-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

67846-500MG 67846-1G 67846-25G 67846-5G
77333-020EA 292.52 USD
77333-020 77357-240 77390-172 77381-150
5-Bromo-2-fluoro-3,4'-dimethoxy-1,1'-biphenyl ≥97%
5-Bromo-2-fluoro-3,4'-dimethoxy-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₂BrFO₂
MW: 311.146 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763041
CAS Number: 2807464-40-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR