5-Bromo-2,4'-difluoro-3-methoxy-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

67844-500MG 67844-1G 67844-5G 67844-25G
77333-016EA 281.19 USD
77333-016 77357-236 77369-048 77390-168
5-Bromo-2,4'-difluoro-3-methoxy-1,1'-biphenyl ≥97%
5-Bromo-2,4'-difluoro-3-methoxy-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₃H₉BrF₂O
Storage Temperature: Ambient
MDL Number: MFCD34763039
CAS Number: 2807442-87-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR