1-Bromo-3-fluoro-2-methoxy-4-(trifluoromethyl)benzene ≥95%

Supplier: AOBChem USA

Warning

67832-500MG 67832-1G 67832-5G 67832-25G
77333-012EA 100.94 USD
77333-012 77342-962 77369-044 77381-148
1-Bromo-3-fluoro-2-methoxy-4-(trifluoromethyl)benzene ≥95%
1-Bromo-3-fluoro-2-methoxy-4-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₈H₅BrF₄O
MW: 273.022 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763033
CAS Number: 2091605-25-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR