2-Fluoro-5-(methylthio)-3-(trifluoromethyl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

67820-500MG 67820-1G 67820-5G
77333-010EA 328.57 USD
77333-010 77357-232 77381-146
2-Fluoro-5-(methylthio)-3-(trifluoromethyl)benzaldehyde ≥97%
2-Fluoro-5-(methylthio)-3-(trifluoromethyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₉H₆F₄OS
MW: 238.202 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763030
CAS Number: 2918895-03-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR