5-Bromo-2-fluoro-1-iodo-3-(trifluoromethyl)benzene ≥97%
Supplier: AOBChem USA
|
Warning
|
67811-500MG
67811-1G
67811-5G
67811-25G
77333-008EA
95.79
USD
77333-008
77342-588
77369-042
77381-144
5-Bromo-2-fluoro-1-iodo-3-(trifluoromethyl)benzene ≥97%
5-Bromo-2-fluoro-1-iodo-3-(trifluoromethyl)benzene
Precaución: for Research Use Only
Formula:
C₇H₂BrF₄I MW: 368.893 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD34763028 CAS Number: 2386270-92-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...