5-Chloro-2-fluoro-1-iodo-3-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

67806-500MG 67806-1G 67806-5G 67806-25G
77333-006EA 114.33 USD
77333-006 77343-370 77369-040 77381-142
5-Chloro-2-fluoro-1-iodo-3-(trifluoromethyl)benzene ≥97%
5-Chloro-2-fluoro-1-iodo-3-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₇H₂ClF₄I
MW: 324.442 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763025
CAS Number: 2384699-46-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR