(5-Chloro-2-fluoro-3-(trifluoromethyl)phenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

67805-500MG 67805-1G 67805-5G 67805-25G
77333-004EA 184.37 USD
77333-004 77347-310 77369-038 77390-164
(5-Chloro-2-fluoro-3-(trifluoromethyl)phenyl)(methyl)sulfane ≥97%
(5-Chloro-2-fluoro-3-(trifluoromethyl)phenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₈H₅ClF₄S
Storage Temperature: Ambient
MDL Number: MFCD34763024
CAS Number: 2705759-95-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR