4'-Phenoxy-[1,1'-biphenyl]-4-carbaldehyde ≥95%

Supplier: AOBChem USA

Warning

67782-500MG 67782-1G 67782-5G
77333-002EA 317.24 USD
77333-002 77357-230 77381-140
4'-Phenoxy-[1,1'-biphenyl]-4-carbaldehyde ≥95%
4'-Phenoxy-[1,1'-biphenyl]-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₉H₁₄O₂
MW: 274.313 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763007
CAS Number: 1406541-79-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR