4'Isopropoxy-[1,1'-biphenyl]-4-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

67775-500MG 67775-1G 67775-25G 67775-5G
77333-000EA 285.31 USD
77333-000 77357-226 77390-162 77381-136
4'Isopropoxy-[1,1'-biphenyl]-4-carbaldehyde ≥97%
4'Isopropoxy-[1,1'-biphenyl]-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₆H₁₆O₂
MW: 240.297 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11919741
CAS Number: 1095989-15-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR