(5-(Trifluoromethyl)-[1,1'-biphenyl]-2-yl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

67752-500MG 67752-1G 67752-5G 67752-25G
77332-992EA 104.03 USD
77332-992 77343-078 77369-030 77381-128
(5-(Trifluoromethyl)-[1,1'-biphenyl]-2-yl)boronic acid ≥97%
(5-(Trifluoromethyl)-[1,1'-biphenyl]-2-yl)boronic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₀BF₃O₂
MW: 266.024 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29067562
CAS Number: 1817781-37-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR