2-Bromo-5-(trifluoromethyl)-1,1'-biphenyl ≥95%

Supplier: AOBChem USA

Warning

67751-1G 67751-5G 67751-25G
77340-804EA 60.77 USD
77340-804 77369-028 77381-126
2-Bromo-5-(trifluoromethyl)-1,1'-biphenyl ≥95%
2-Bromo-5-(trifluoromethyl)-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₃H₈BrF₃
MW: 301.102 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762985
CAS Number: 2096967-58-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR