2-Methoxy-5-phenoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

67733-500MG 67733-1G 67733-5G 67733-25G
77332-990EA 176.13 USD
77332-990 77346-546 77369-026 77390-156
2-Methoxy-5-phenoxybenzoic acid ≥97%
2-Methoxy-5-phenoxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₂O₄
MW: 244.243 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19309485
CAS Number: 63987-24-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR