2-Bromo-4,5-difluoro-4'-(trifluoromethoxy)-1,1'-biphenyl ≥95%
Supplier: AOBChem USA
|
Warning
|
67723-500MG
67723-1G
67723-5G
67723-25G
77332-986EA
281.19
USD
77332-986
77357-222
77369-022
77390-152
2-Bromo-4,5-difluoro-4'-(trifluoromethoxy)-1,1'-biphenyl ≥95%
2-Bromo-4,5-difluoro-4'-(trifluoromethoxy)-1,1'-biphenyl
Precaución: for Research Use Only
Formula:
C₁₃H₆BrF₅O MW: 353.082 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD34762979 CAS Number: 2901102-35-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...