2-Bromo-4-fluoro-3'-methoxy-6-methyl-1,1'-biphenyl ≥95%

Supplier: AOBChem USA

Warning

67718-500MG 67718-1G 67718-5G 67718-25G
77332-984EA 188.49 USD
77332-984 77357-220 77369-020 77390-150
2-Bromo-4-fluoro-3'-methoxy-6-methyl-1,1'-biphenyl ≥95%
2-Bromo-4-fluoro-3'-methoxy-6-methyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₂BrFO
MW: 295.147 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762976
CAS Number: 2901102-46-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR