2-Bromo-4-fluoro-6-methyl-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

67696-500MG 67696-1G 67696-5G
77336-016EA 373.89 USD
77336-016 77357-214 77381-122
2-Bromo-4-fluoro-6-methyl-1,1'-biphenyl ≥97%
2-Bromo-4-fluoro-6-methyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₃H₁₀BrF
MW: 265.121 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762957
CAS Number: 2901066-24-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR