4'-Isopropoxy-[1,1'-biphenyl]-4-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

67667-1G 67667-5G 67667-25G
77342-032EA 111.24 USD
77342-032 77369-016 77381-120
4'-Isopropoxy-[1,1'-biphenyl]-4-carboxylic acid ≥97%
4'-Isopropoxy-[1,1'-biphenyl]-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₆H₁₆O₃
MW: 256.296 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11923479
CAS Number: 502649-35-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR