4-Fluoro-4'-(trifluoromethoxy)-3-(trifluoromethyl)-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

67644-500MG 67644-1G 67644-5G 67644-25G
77332-974EA 184.37 USD
77332-974 77347-308 77369-006 77390-140
4-Fluoro-4'-(trifluoromethoxy)-3-(trifluoromethyl)-1,1'-biphenyl ≥97%
4-Fluoro-4'-(trifluoromethoxy)-3-(trifluoromethyl)-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₇F₇O
MW: 324.194 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18406645
CAS Number: 1261655-93-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR