4-Fluoro-3-iodo-5-(trifluoromethyl)-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

67632-500MG 67632-1G 67632-5G 67632-25G
77332-970EA 231.75 USD
77332-970 77357-206 77369-002 77390-136
4-Fluoro-3-iodo-5-(trifluoromethyl)-1,1'-biphenyl ≥97%
4-Fluoro-3-iodo-5-(trifluoromethyl)-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₃H₇F₄I
MW: 366.093 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762920
CAS Number: 2901098-52-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR