4-Fluoro-5-(trifluoromethyl)-[1,1'-biphenyl]-3-carboxylic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
67628-500MG
67628-1G
67628-5G
67628-25G
77332-968EA
180.25
USD
77332-968
77346-762
77369-000
77390-134
4-Fluoro-5-(trifluoromethyl)-[1,1'-biphenyl]-3-carboxylic acid ≥97%
4-Fluoro-5-(trifluoromethyl)-[1,1'-biphenyl]-3-carboxylic acid
Precaución: for Research Use Only
Formula:
C₁₄H₈F₄O₂ MW: 284.206 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD34762916 CAS Number: 2901066-67-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...