2-Fluoro-5-iodo-3-(trifluoromethyl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

67621-1G 67621-5G 67621-25G
77341-358EA 82.4 USD
77341-358 77368-998 77381-116
2-Fluoro-5-iodo-3-(trifluoromethyl)benzaldehyde ≥97%
2-Fluoro-5-iodo-3-(trifluoromethyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₈H₃F₄IO
Storage Temperature: Ambient
MDL Number: MFCD34762911
CAS Number: 2090966-85-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR