2-(Benzyloxy)-3-bromo-5-(trifluoromethyl)benzoic acid ≥95%

Supplier: AOBChem USA

Warning

67576-500MG 67576-1G 67576-5G
77335-900EA 367.71 USD
77335-900 77357-202 77381-108
2-(Benzyloxy)-3-bromo-5-(trifluoromethyl)benzoic acid ≥95%
2-(Benzyloxy)-3-bromo-5-(trifluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₀BrF₃O₃
MW: 375.137 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762890
CAS Number: 2807443-15-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR