3-Bromo-2-ethoxy-5-(trifluoromethyl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

67571-500MG 67571-1G 67571-5G 67571-25G
77332-962EA 272.95 USD
77332-962 77357-200 77368-992 77390-132
3-Bromo-2-ethoxy-5-(trifluoromethyl)benzaldehyde ≥97%
3-Bromo-2-ethoxy-5-(trifluoromethyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₈BrF₃O₂
MW: 297.069 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762886
CAS Number: 2387200-43-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR