3-Bromo-2-ethoxy-5-(trifluoromethyl)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

67570-500MG 67570-1G 67570-5G
77335-898EA 367.71 USD
77335-898 77357-198 77381-106
3-Bromo-2-ethoxy-5-(trifluoromethyl)benzoic acid ≥97%
3-Bromo-2-ethoxy-5-(trifluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₈BrF₃O₃
MW: 313.068 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762885
CAS Number: 2701484-35-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR