3-Iodo-5-methoxybenzonitrile ≥97%

Supplier: AOBChem USA

Warning

67567-500MG 67567-1G 67567-5G
77332-958EA 326.51 USD
77332-958 77357-196 77381-104
3-Iodo-5-methoxybenzonitrile ≥97%
3-Iodo-5-methoxybenzonitrile

Precaución: for Research Use Only

Formula: C₈H₆INO
Storage Temperature: Ambient
MDL Number: MFCD12828564
CAS Number: 269411-72-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR