1-(4-Bromo-1-fluoronaphthalen-2-yl)-2,2-difluoroethanone ≥97%

Supplier: AOBChem USA

Warning

67554-500MG 67554-1G 67554-5G 67554-25G
77332-954EA 202.91 USD
77332-954 77357-190 77368-988 77390-128
1-(4-Bromo-1-fluoronaphthalen-2-yl)-2,2-difluoroethanone ≥97%
1-(4-Bromo-1-fluoronaphthalen-2-yl)-2,2-difluoroethanone

Precaución: for Research Use Only

Formula: C₁₂H₆BrF₃O
MW: 303.075 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762878
CAS Number: 2807450-76-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR