1-(4-Bromo-1-fluoronaphthalen-2-yl)-2,2,2-trifluoroethanone ≥97%

Supplier: AOBChem USA

Warning

67553-500MG 67553-1G 67553-5G 67553-25G
77332-952EA 204.97 USD
77332-952 77357-188 77368-986 77390-126
1-(4-Bromo-1-fluoronaphthalen-2-yl)-2,2,2-trifluoroethanone ≥97%
1-(4-Bromo-1-fluoronaphthalen-2-yl)-2,2,2-trifluoroethanone

Precaución: for Research Use Only

Formula: C₁₂H₅BrF₄O
MW: 321.065 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762877
CAS Number: 2807447-29-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR