3-Bromo-2-chloro-6-fluoro-5-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

67550-1G 67550-5G 67550-25G
77341-834EA 105.06 USD
77341-834 77368-984 77381-100
3-Bromo-2-chloro-6-fluoro-5-methylbenzoic acid ≥97%
3-Bromo-2-chloro-6-fluoro-5-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₅BrClFO₂
Storage Temperature: Ambient
MDL Number: MFCD34762875
CAS Number: 2807464-77-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR