1-(3-Bromo-2-chloro-6-fluoro-5-methylphenyl)-2,2,2-trifluoroethanone ≥97%

Supplier: AOBChem USA

Warning

67549-500MG 67549-1G 67549-5G 67549-25G
77332-950EA 99.91 USD
77332-950 77342-872 77368-982 77381-098
1-(3-Bromo-2-chloro-6-fluoro-5-methylphenyl)-2,2,2-trifluoroethanone ≥97%
1-(3-Bromo-2-chloro-6-fluoro-5-methylphenyl)-2,2,2-trifluoroethanone

Precaución: for Research Use Only

Formula: C₉H₄BrClF₄O
MW: 319.478 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762874
CAS Number: 2807440-12-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR