2-Bromo-3-chloro-6-fluoro-5-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

67545-500MG 67545-1G 67545-5G 67545-25G
77332-948EA 136.99 USD
77332-948 77344-626 77368-980 77390-124
2-Bromo-3-chloro-6-fluoro-5-methylbenzoic acid ≥97%
2-Bromo-3-chloro-6-fluoro-5-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₅BrClFO₂
Storage Temperature: Ambient
MDL Number: MFCD34754966
CAS Number: 2807454-56-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR