2-Bromo-3-chloro-6-fluoro-5-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

67544-500MG 67544-1G 67544-5G 67544-25G
77332-946EA 178.19 USD
77332-946 77346-620 77368-978 77390-122
2-Bromo-3-chloro-6-fluoro-5-methylbenzaldehyde ≥97%
2-Bromo-3-chloro-6-fluoro-5-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₅BrClFO
Storage Temperature: Ambient
MDL Number: MFCD34754997
CAS Number: 2807443-00-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR