4-Bromo-2,6-difluoro-3-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

67541-500MG 67541-1G 67541-5G 67541-25G
77332-944EA 116.39 USD
77332-944 77343-454 77368-976 77381-096
4-Bromo-2,6-difluoro-3-methoxybenzoic acid ≥97%
4-Bromo-2,6-difluoro-3-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₈H₅BrF₂O₃
MW: 267.024 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762872
CAS Number: 2807440-30-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR