4-Bromo-2,6-difluoro-3-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

67540-500MG 67540-1G 67540-5G 67540-25G
77332-942EA 99.91 USD
77332-942 77342-870 77368-974 77381-094
4-Bromo-2,6-difluoro-3-methoxybenzaldehyde ≥97%
4-Bromo-2,6-difluoro-3-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₅BrF₂O₂
MW: 251.025 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762871
CAS Number: 2807445-60-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR