1-(4-Chloro-3-methoxyphenyl)-2,2-difluoroethanone ≥97%

Supplier: AOBChem USA

Warning

67509-500MG 67509-1G 67509-5G 67509-25G
77332-930EA 114.33 USD
77332-930 77343-368 77368-964 77381-084
1-(4-Chloro-3-methoxyphenyl)-2,2-difluoroethanone ≥97%
1-(4-Chloro-3-methoxyphenyl)-2,2-difluoroethanone

Precaución: for Research Use Only

Formula: C₉H₇ClF₂O₂
MW: 220.6 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26045105
CAS Number: 1565686-07-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR