1-(4-Chloro-3-ethoxyphenyl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

67507-500MG 67507-1G 67507-5G 67507-25G
77332-928EA 143.17 USD
77332-928 77345-008 77368-962 77390-110
1-(4-Chloro-3-ethoxyphenyl)-2,2-difluoroethanone ≥95%
1-(4-Chloro-3-ethoxyphenyl)-2,2-difluoroethanone

Precaución: for Research Use Only

Formula: C₁₀H₉ClF₂O₂
MW: 234.627 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762855
CAS Number: 2807450-71-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR