5-Bromo-2-fluoro-1-methyl-3-(methylsulfonyl)benzene ≥97%

Supplier: AOBChem USA

Warning

67503-500MG 67503-1G 67503-5G 67503-25G
77332-924EA 157.59 USD
77332-924 77345-714 77368-958 77390-106
5-Bromo-2-fluoro-1-methyl-3-(methylsulfonyl)benzene ≥97%
5-Bromo-2-fluoro-1-methyl-3-(methylsulfonyl)benzene

Precaución: for Research Use Only

Formula: C₈H₈BrFO₂S
MW: 267.115 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762852
CAS Number: 2807442-76-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR