(4-Fluoro-5-methyl-1,3-phenylene)bis(methylsulfane) ≥97%

Supplier: AOBChem USA

Warning

67502-500MG 67502-1G 67502-5G
77335-610EA 360.5 USD
77335-610 77357-174 77381-082
(4-Fluoro-5-methyl-1,3-phenylene)bis(methylsulfane) ≥97%
(4-Fluoro-5-methyl-1,3-phenylene)bis(methylsulfane)

Precaución: for Research Use Only

Formula: C₉H₁₁FS₂
MW: 202.312 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762851
CAS Number: 2807456-05-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR