(3-Bromo-2-fluoro-5-(trifluoromethyl)phenyl)trimethylsilane ≥97%

Supplier: AOBChem USA

Warning

67491-500MG 67491-1G 67491-5G 67491-25G
77332-916EA 157.59 USD
77332-916 77345-712 77368-952 77390-100
(3-Bromo-2-fluoro-5-(trifluoromethyl)phenyl)trimethylsilane ≥97%
(3-Bromo-2-fluoro-5-(trifluoromethyl)phenyl)trimethylsilane

Precaución: for Research Use Only

Formula: C₁₀H₁₁BrF₄Si
MW: 315.177 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762843
CAS Number: 2807471-02-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR