3,5-Dibromo-N,N-dimethylaniline ≥97%

Supplier: AOBChem USA

Warning

67489-500MG 67489-1G 67489-5G 67489-25G
77332-914EA 184.37 USD
77332-914 77347-304 77368-950 77390-098
3,5-Dibromo-N,N-dimethylaniline ≥97%
3,5-Dibromo-N,N-dimethylaniline

Precaución: for Research Use Only

Formula: C₈H₉Br₂N
MW: 278.972 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22042773
CAS Number: 64230-29-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR