3-Bromo-5-fluoro-4'-(trifluoromethoxy)-1,1'-biphenyl ≥97%
Supplier: AOBChem USA
|
Warning
|
67488-500MG
67488-1G
67488-5G
67488-25G
77332-912EA
141.11
USD
77332-912
77344-880
77368-948
77390-096
3-Bromo-5-fluoro-4'-(trifluoromethoxy)-1,1'-biphenyl ≥97%
3-Bromo-5-fluoro-4'-(trifluoromethoxy)-1,1'-biphenyl
Precaución: for Research Use Only
Formula:
C₁₃H₇BrF₄O MW: 335.092 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD34762842 CAS Number: 2807440-51-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...