3-Bromo-4',5-difluoro-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

67474-500MG 67474-1G 67474-5G 67474-25G
77332-902EA 99.91 USD
77332-902 77342-868 77368-944 77381-070
3-Bromo-4',5-difluoro-1,1'-biphenyl ≥97%
3-Bromo-4',5-difluoro-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₂H₇BrF₂
MW: 269.085 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762834
CAS Number: 160521-37-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR