3'-Bromo-5'-fluoro-2-methyl-1,1'-biphenyl ≥95%

Supplier: AOBChem USA

Warning

67473-500MG 67473-1G 67473-5G 67473-25G
77332-900EA 120.51 USD
77332-900 77343-798 77368-942 77381-068
3'-Bromo-5'-fluoro-2-methyl-1,1'-biphenyl ≥95%
3'-Bromo-5'-fluoro-2-methyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₃H₁₀BrF
Storage Temperature: Ambient
MDL Number: MFCD34762833
CAS Number: 2807471-77-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR