3-Bromo-5-fluoro-3'-methyl-1,1'-biphenyl ≥95%

Supplier: AOBChem USA

Warning

67472-500MG 67472-1G 67472-5G 67472-25G
77332-898EA 120.51 USD
77332-898 77343-796 77368-940 77381-066
3-Bromo-5-fluoro-3'-methyl-1,1'-biphenyl ≥95%
3-Bromo-5-fluoro-3'-methyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₃H₁₀BrF
Storage Temperature: Ambient
MDL Number: MFCD34762832
CAS Number: 2807470-45-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR